Learn More
OspC Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | OspC |
---|---|
Concentration | 0.95 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-194-3808
![]() ![]() |
Rockland Immunochemicals
200401C11 |
100 μg |
Each for $636.24
|
|
|||||
50-105-7638
![]() ![]() |
Rockland Immunochemicals
200401C11S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for Borrelia burgdorferi OspC protein. A BLAST analysis was used to suggest cross-reactivity with p39 from B. burgdorferi, afzelii, and valaisiana sources based on 100% homology with the immunizing sequence. Partial reactivity is expected against B. japonica and americana sources based on 94% homology. Cross-reactivity with OspC from other sources has not been determined.
Outer Surface Protein C, or OspC, is a 20.7 kDa immunogenic protein on the outer surface of the spirochete Borrelia burgdorferi. Its function is not known, but it is located with lipid-anchoring sites on the outer cell membrane.Specifications
OspC | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Bacteria | |
MBP-fusion protein corresponding to Borrelia burgdorferi OspC protein. | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.95 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.