Learn More
OSR1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | OSR1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2296
![]() |
Bethyl Laboratories, Inc
IHC00552T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2295
![]() |
Bethyl Laboratories, Inc
IHC00552 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine, Dog, Horse, Guinea pig_10141, Orangutan, Monkey, Gorilla, Chimpanzee, Chine
Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.Specifications
OSR1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
OTTHUMP00000209062; oxidative stress-responsive 1 protein; serine/threonine-protein kinase OSR1 | |
OXSR1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O95747 | |
9943 | |
Between 375 and 425 | |
Primary | |
4° C | |
OXSR1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.