Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoactivin/GPNMB Antibody (303822), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB364781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364781 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364781 Supplier Novus Biologicals Supplier No. FAB25501X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteoactivin/GPNMB Monoclonal antibody specifically detects Osteoactivin/GPNMB in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen Osteoactivin/GPNMB
Applications Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 303822
Conjugate Alexa Fluor 532
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias glycoprotein (transmembrane) nmb, glycoprotein NMB, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Osteoactivin/GPNMB isoform 2, Ala22-Asn486, Accession # NP_002501
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10457
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.