Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoadherin/OSAD/OMD Antibody (107), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB075427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075427 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075427 Supplier Novus Biologicals Supplier No. NBP290596AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Osteoadherin/OSAD/OMD Monoclonal specifically detects Osteoadherin/OSAD/OMD in Mouse samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen Osteoadherin/OSAD/OMD
Applications ELISA
Classification Monoclonal
Clone 107
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2Costeoadherin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Osteoadherin/OSAD/OMD (NP_036180.1; Met1-Ile423).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4958
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.