Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteocalcin Antibody (2H9F11F8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062801 0.1 mg
NB017511 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062801 Supplier Novus Biologicals Supplier No. NB10062801
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteocalcin Monoclonal specifically detects Osteocalcin in Human, Mouse (Negative), Rat samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Osteocalcin
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2H9F11F8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry
Gene Accession No. P02818
Gene Alias BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin
Gene Symbols BGLAP
Host Species Mouse
Immunogen Recombinant human osteocalcin protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 632
Test Specificity This product recognizes intact human and bovine osteocalcin in a “2 site” assay and reacts with osteocalcin fragments corresponding to amino acids 15-31 and 20-43 of the native molecule. It does not recognize fragments representing amino acids 1-19 or 7-19.
Target Species Human, Rat, Mouse (Negative), Rat, Mouse (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.