Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteopontin Mouse anti-Human,Mouse, Unconjugated, Clone: 2A1, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Osteopontin |
---|---|
Clone | 2A1 |
Applications | ELISA, Immunohistochemistry, Affinity Purification, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15172MI
|
MilliporeSigma
MABT151725UG |
25 μg |
Each for $182.31
|
|
|||||
MABT15171MI
|
MilliporeSigma
MABT1517100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
Osteopontin Monoclonal antibody specifically detects Osteopontin in Human,Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoaffinity Purification, Immunoprecipitation, Western Blotting.Specifications
Osteopontin | |
ELISA, Immunohistochemistry, Affinity Purification, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Bone sialoprotein 1;Nephropontin;Secreted phosphoprotein 1;SPP-1;Urinary stone protein;OPN;Uropontin | |
His-tagged full-length recombinant human Osteopontin. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼26 kDa observed; 35.42 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
2A1 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_000573 | |
SPP1, BNSP, OPN, PSEC0156 | |
IgG1 κ | |
Protein G Purified | |
Clone 2A1 is a mouse monoclonal antibody that specifically detects Osteopontin. It targets an epitope within the C-terminal half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title