Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteopontin/OPN Antibody (1B20), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB11089062C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11089062C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11089062C Supplier Novus Biologicals Supplier No. NB11089062C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Osteopontin/OPN
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1B20
Conjugate DyLight 650
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P10451
Gene Alias BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Gene Symbols SPP1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminus of human Osteopontin [Swiss-Prot# P10451].
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 6696
Target Species Human, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.