Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteopontin/OPN Antibody, Novus Biologicals™
SDP

Catalog No. NB600104301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600104301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600104301 Supplier Novus Biologicals Supplier No. NB60010430.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

Osteopontin/OPN Polyclonal specifically detects Osteopontin/OPN in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Osteopontin/OPN
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:20000, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Accession No. p10451
Gene Alias BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Gene Symbols SPP1
Host Species Rabbit
Immunogen This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide, from the human osteopontin protein, conjugated to KLH using maleimide.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6696
Test Specificity This recognises the full length osteopontin protein (which runs at 66 kD on westerns), as well as the C-terminal fragments of both thrombin and MMP-cleaved OPN. This recognises the 32 kD MMP-cleaved C-fragment, but not the 40 kD N-terminal fragment.
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.