Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin/TNFRSF11B Antibody (155321), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311045 Supplier Novus Biologicals Supplier No. FAB8052PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications Flow Cytometry
Classification Monoclonal
Clone 155321
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B, Glu22-Leu401, Accession # AAB53709.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4982
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.