Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin/TNFRSF11B Antibody (340), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB307843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307843 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307843 Supplier Novus Biologicals Supplier No. NBP289371MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications ELISA
Classification Monoclonal
Clone 340
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoprotegerin/TNFRSF11B (Accession#: NP_002537.3; Met 1-Leu 401).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4982
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.