Learn More
Osteoprotegerin/TNFRSF11B Antibody (98A1071) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Osteoprotegerin/TNFRSF11B |
| Applications | CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 98A1071 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/ml, Flow Cytometry 0.2-1 ug/1 million cells, ELISA reported in scientific literature (PMID 15935726), Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000. Use reported in scientific literature (Sandra (2006)), Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Sandra (2006)), Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Boumans (2012)), CyTOF-ready |
| Gene Accession No. | O00300 |
| Gene Alias | OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b |
| Gene Symbols | TNFRSF11B |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.