Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin/TNFRSF11B Antibody (98A1071) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056505T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056505T 0.025 mg
NB10056505 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056505T Supplier Novus Biologicals Supplier No. NB10056505SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 35 publications

Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Guinea Pig, Human, Mouse, Rabbit, Rat samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 98A1071
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Flow Cytometry 0.2-1 ug/1 million cells, ELISA reported in scientific literature (PMID 15935726), Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000. Use reported in scientific literature (Sandra (2006)), Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Sandra (2006)), Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Boumans (2012)), CyTOF-ready
Gene Accession No. O00300
Gene Alias OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Gene Symbols TNFRSF11B
Host Species Mouse
Immunogen A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 4982
Test Specificity The Osteoprotegerin antibody, clone 98A1071, detects all forms of Osteoprotegerin including monomeric, dimeric and Osteoprotegerin bound to RANKL (Vandooren, 2008) as well as Osteoprotegerin bound to TRAIL (Sandra, 2006).
Target Species Guinea Pig, Mouse, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.