Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin/TNFRSF11B Antibody (98A1071), PerCP, Novus Biologicals™
SDP

Catalog No. NB10056505P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056505P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056505P Supplier Novus Biologicals Supplier No. NB10056505PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human, Mouse, Rabbit, Rat samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 98A1071
Conjugate PerCP
Dilution Flow Cytometry 1 μL/1 million cells
Gene Accession No. O00300
Gene Alias OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Gene Symbols TNFRSF11B
Host Species Mouse
Immunogen A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4982
Test Specificity The Osteoprotegerin antibody, clone 98A1071, detects all forms of Osteoprotegerin including monomeric, dimeric and Osteoprotegerin bound to RANKL (Vandooren, 2008) as well as Osteoprotegerin bound to TRAIL (Sandra, 2006).
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.