Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osterix/Sp7 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB320233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320233 Supplier Novus Biologicals Supplier No. NBP298576F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Osterix/Sp7 Polyclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot

Specifications

Antigen Osterix/Sp7
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias MGC126598, OSXosterix, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Osterix/Sp7 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121340
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.