Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSTF1 Mouse anti-Human, Clone: 9G4, Novus Biologicals™
SDP

Catalog No. NBP25939650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25939650 50 μL
NBP259396X 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25939650 Supplier Novus Biologicals Supplier No. NBP25939650UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSTF1 Monoclonal specifically detects OSTF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OSTF1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9G4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1009, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1
Gene Symbols OSTF1
Host Species Mouse
Immunogen Recombinant human OSTF1 (aa) purified from E. coli
Purification Method Protein G purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26578
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.