Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Otubain-1 Antibody (17E10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309089 Supplier Novus Biologicals Supplier No. NBP259389MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Otubain-1 Monoclonal antibody specifically detects Otubain-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Otubain-1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 17E10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1
Host Species Mouse
Immunogen Recombinant human Otubain-1 (1-271aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 55611
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.