Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Otubain-2 Antibody (139), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB038034 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038034 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038034 Supplier Novus Biologicals Supplier No. NBP290191C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Otubain-2 Monoclonal specifically detects Otubain-2 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Otubain-2
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 139
Conjugate DyLight 650
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Otubain-2 (Q96DC9-1; Met1-His234).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78990
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.