Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD4 Antibody (3B12G11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252525 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252525 Supplier Novus Biologicals Supplier No. NBP252525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OTUD4 Monoclonal specifically detects OTUD4 in Human, Rat samples. It is validated for CyTOF, ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen OTUD4
Applications CyTOF, ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3B12G11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4
Gene Symbols OTUD4
Host Species Mouse
Immunogen Purified recombinant fragment of human OTUD4 (AA: 815-1049) expressed in E. Coli.
Molecular Weight of Antigen 124 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54726
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.