Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUD7B Mouse anti-Human, Clone: 2C12D9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | OTUD7B |
---|---|
Clone | 2C12D9 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6708
![]() ![]() |
Proteintech Group Inc
662761IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Negative regulator of the non-canonical NF-kappa-B pathway that acts by mediating deubiquitination of TRAF3, an inhibitor of the NF-kappa-B pathway, thereby acting as a negative regulator of B-cell responses. In response to non-canonical NF- kappa-B stimuli, deubiquitinates ′Lys-48′-linked polyubiquitin chains of TRAF3, preventing TRAF3 proteolysis and over-activation of non-canonical NF-kappa-B. Negatively regulates mucosal immunity against infections. Mediates deubiquitination of EGFR. Has deubiquitinating activity toward ′Lys-11′, ′Lys-48′ or ′Lys-63′- linked polyubiquitin chains. In vitro, has preference for ′Lys- 11′-linked polyubiquitin chains; however such data are unsure in vivo. Hydrolyzes both linear and branched forms of polyubiquitin.Specifications
OTUD7B | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CEZANNE, OTU domain containing 7B, OTUD7B, ZA20D1, Zinc finger protein Cezanne | |
OTUD7B | |
IgG2b | |
Protein A | |
Antibody |
2C12D9 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q6GQQ9 | |
56957 | |
OTUD7B Fusion Protein Ag10513 | |
Primary | |
-20°C | |
OTUD7B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title