Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Otx1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB019224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019224 0.025 mg
NBP212545 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019224 Supplier Novus Biologicals Supplier No. NBP2125450.025MG

Rabbit Polyclonal Antibody

Otx1 Polyclonal specifically detects Otx1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Otx1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P32242
Gene Alias FLJ38361, homeobox protein OTX1, MGC15736, orthodenticle (Drosophila) homolog 1, orthodenticle homeobox 1, Orthodenticle homolog 1, orthodenticle homolog 1 (Drosophila)
Gene Symbols OTX1
Host Species Rabbit
Immunogen Antibody target: OTX1 KLH conjugated synthetic peptide between 86-116 amino acids from the Central region of human OTX1.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5013
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.