Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTX3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18025520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18025520 20 μL
NBP180255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18025520 Supplier Novus Biologicals Supplier No. NBP18025520UL

Rabbit Polyclonal Antibody

OTX3 Polyclonal specifically detects OTX3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen OTX3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_001020738
Gene Alias diencephalon/mesencephalon homeobox 1, diencephalon/mesencephalon homeobox protein 1, homeoprotein MBX, MBX, Orthodenticle homolog 3, OTX3, Paired-like homeobox protein DMBX1, PAXBorthodenticle homolog 3 (Drosophila)
Gene Symbols DMBX1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of mouse DMBX1. Peptide sequence LSLSEQSASESAPEDQLDREEDSRAEEPKAEKSPGSESKVPGCKRGSPKA.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127343
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.