Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OV-6 Antibody (OV-6), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB366686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366686 Supplier Novus Biologicals Supplier No. FAB2020M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OV-6 Monoclonal specifically detects OV-6 in Human, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen OV-6
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone OV-6
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Carcinogen-treated rat nodular hepatocytes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.