Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval Cell Marker Antibody (OC2-3C7), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP118970A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP118970A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP118970A70 Supplier Novus Biologicals Supplier No. NBP118970AF750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Oval Cell Marker
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OC2-3C7
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Rat
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for both proliferating and normal hepatic periductal cells.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.