Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oval Cell Marker Antibody (OC2-4E8), Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP118971SS
Description
Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Oval Cell Marker | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Frozen 1:100 | |
macrophage inhibitory cytokine-1 | |
Nonparenchymal cells from DDC-treated mice to induce the oval cell response. | |
0.025 mL | |
Hepatic Stem Cell Markers, Stem Cell Markers | |
Specific for both proliferating and normal hepatic periductal cells. | |
Store at 4C. Do not freeze. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
OC2-4E8 | |
Unconjugated | |
Tissue culture supernatant with 0.05% Sodium Azide | |
Rat | |
Tissue culture supernatant | |
RUO | |
Primary | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction