Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oval Cell Marker Antibody (OC2-4E8), Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | Oval Cell Marker |
---|---|
Clone | OC2-4E8 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Frozen 1:100 |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP118971SS
![]() |
Novus Biologicals
NBP118971SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP118971
![]() |
Novus Biologicals
NBP118971 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Oval Cell Marker | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Hepatic Stem Cell Markers, Stem Cell Markers | |
Tissue culture supernatant with 0.05% Sodium Azide | |
Nonparenchymal cells from DDC-treated mice to induce the oval cell response. | |
Primary | |
Store at 4C. Do not freeze. |
OC2-4E8 | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Frozen 1:100 | |
Monoclonal | |
Supernatant | |
RUO | |
Mouse | |
macrophage inhibitory cytokine-1 | |
IgG2b | |
Tissue culture supernatant | |
Specific for both proliferating and normal hepatic periductal cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title