Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval Cell Marker Antibody (OC2-6E10), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N118981PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N118981PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N118981PEC7 Supplier Novus Biologicals Supplier No. NBP118981PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Oval Cell Marker
Applications Flow Cytometry
Classification Monoclonal
Clone OC2-6E10
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Rat
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for proliferating hepatic periductal cells.
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.