Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Antibody (2C6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062809
Description
Ovalbumin Monoclonal specifically detects Ovalbumin in Chicken samples. It is validated for ELISA.Specifications
Ovalbumin | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:1000-1:5000 | |
OVAL | |
Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma, P3U1 | |
0.1 mg | |
Primary | |
NB100-62809 is a mouse monoclonal antibody that specifically recognises ovalbumin (OVA). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgE |
ELISA | |
2C6 | |
Unconjugated | |
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
Mouse | |
Protein G purified | |
RUO | |
396058 | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction