Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Antibody (2C6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Ovalbumin |
---|---|
Clone | 2C6 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:1000-1:5000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017514
![]() |
Novus Biologicals
NB100628090.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10062809
![]() |
Novus Biologicals
NB10062809 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Ovalbumin Monoclonal specifically detects Ovalbumin in Chicken samples. It is validated for ELISA.Specifications
Ovalbumin | |
1.0 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Chicken | |
396058 | |
Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma, P3U1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2C6 | |
ELISA 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
OVAL | |
IgE | |
Protein G purified | |
NB100-62809 is a mouse monoclonal antibody that specifically recognises ovalbumin (OVA). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title