Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin Antibody (2C6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10062809C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062809C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062809C Supplier Novus Biologicals Supplier No. NB10062809C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ovalbumin Monoclonal specifically detects Ovalbumin in Chicken samples. It is validated for ELISA.

Specifications

Antigen Ovalbumin
Applications ELISA
Classification Monoclonal
Clone 2C6
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
Gene Symbols OVAL
Host Species Mouse
Immunogen Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma, P3U1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396058
Test Specificity NB100-62809 is a mouse monoclonal antibody that specifically recognises ovalbumin (OVA).
Target Species Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.