Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ovalbumin Monoclonal Antibody (3#)
GREENER_CHOICE

Catalog No. p-200124257
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541656 200 μL
PIMA543540 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541656 Supplier Invitrogen™ Supplier No. MA541656
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ovalbumin Monoclonal Antibody for Western Blot

Chicken ovalbumin is the major protein in the 'white' of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ovalbumin
Applications Western Blot
Classification Monoclonal
Clone 3#
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene OVAL
Gene Accession No. P01012
Gene Alias Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product
Gene Symbols OVAL
Host Species Mouse
Immunogen Native protein Ovalbumin.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396058
Target Species Multi-species
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.