Learn More
Ovalbumin Polyclonal Antibody, FITC, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | Ovalbumin |
---|---|
Concentration | 1 mg/mL |
Applications | Dot Blot, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-105-7678
![]() ![]() |
Rockland Immunochemicals
20042330100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-1068
![]() ![]() |
Rockland Immunochemicals
2004233S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Ovalbumin Antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-fluorescein, anti-Rabbit Serum as well as purified and partially purified Ovalbumin [Hen Egg White]. Cross reactivity against Ovalbumin from other tissues and species may occur but have not been specifically determined.
Chicken ovalbumin is the major protein in the 'white' of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.Specifications
Ovalbumin | |
Dot Blot, Immunofluorescence, Immunoprecipitation, Western Blot | |
FITC | |
Rabbit | |
Chicken | |
P01012 | |
396058 | |
Ovalbumin [Hen Egg White] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
OVAL |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Allergen Gal d II; clade (ovalbumin); Egg albumin; Gal d 2; member 14; OVAL; Ovalbumin; ovalbumin (SERPINB14); Plakalbumin; serpin peptidase inhibitor; SERPINB14 | |
OVAL | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.