Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Rabbit anti-Chicken, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2524030.1ML
Description
Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA.Specifications
Ovalbumin | |
Polyclonal | |
Unconjugated | |
Phosphate buffered saline with 0.09% sodium azide | |
OVAL | |
Highly purified albumin from chicken egg. | |
Primary | |
Chicken | |
IgG |
Western Blot, ELISA | |
5.0 mg/mL | |
Western Blot 1:4000, ELISA 1:200000 | |
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
Rabbit | |
0.1 mL | |
396058 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction