Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Rabbit anti-Chicken, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
Antigen | Ovalbumin |
---|---|
Concentration | 5.0 mg/mL |
Dilution | Western Blot 1:4000, ELISA 1:200000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019699
![]() |
Novus Biologicals
NBP2524030.1ML |
0.1 mL | N/A | N/A | N/A | ||||
Description
Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA.Specifications
Ovalbumin | |
Western Blot 1:4000, ELISA 1:200000 | |
Polyclonal | |
Rabbit | |
Phosphate buffered saline with 0.09% sodium azide | |
396058 | |
Highly purified albumin from chicken egg. | |
Primary |
5.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Chicken | |
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
OVAL | |
IgG | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title