Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OVGP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357001 Supplier Novus Biologicals Supplier No. NBP176939MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OVGP1 Polyclonal antibody specifically detects OVGP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen OVGP1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CHIT5, EGP, Estrogen-dependent oviduct protein, MUC9, mucin 9, mucin-9, OGP, oviduct glycoprotein, Oviductal glycoprotein, oviductal glycoprotein 1, 120kDa, oviductin, oviduct-specific glycoprotein
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human OVGP1. The immunogen is located within amino acids 60 - 110 of OVGP1. Amino Acid Squence: ILYPEFNKLKERNRELKT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5016
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.