Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ A2 Large Gel Systems

Catalog No. 09528101
Click to view available options
Type:
Large Gel Electrophoresis System
Large, w/Buffer Exchange Ports

Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5mm thick
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Complete system
Product Line Owl
Type Large Gel Electrophoresis System
Sample Size 8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer 2.3 L
Dimensions (L x W x H) 14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
Length (English) 14.8 in.
Length (Metric) 37.5 cm
Length (Metric) Gel 25 cm
Width (English) 12.8 in.
Width (Metric) 32.5 cm
Width (Metric) Gel 20 cm
Height (English) 4.1 in.
Height (Metric) 10.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.