Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Owl™ A2 Large Gel Systems
Click to view available options
Type:
Large Gel Electrophoresis System
Large, w/Buffer Exchange Ports
Description
Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Features:
- Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
- Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
- UVT gel tray has gasketed end gates for leakproof seal without tape
- Built-in leveling ensures flat, uniformly thick gels
- Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
- Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
- Buffer chamber with three-point leveling base
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- UVT gel tray
- Gasketed end gates
- Three combs (16-, 24- and 36-well), 1.5mm thick

Specifications
Specifications
Certifications/Compliance | CE marked |
Description | Complete system with buffer exchange ports |
Product Line | Owl |
Type | Large, w/Buffer Exchange Ports |
Sample Size | 8 to 432 (16-, 24- and 36-well combs) |
Volume (Metric) Buffer | 2.3 L |
Dimensions (L x W x H) | 14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm) |
Length (English) | 14.8 in. |
Length (Metric) | 37.5 cm |
Length (Metric) Gel | 25 cm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction