Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ Aluminum-Backed Sequencers

Catalog No. p-4528732
Click to view available options
Volume (Metric) Buffer:
800mL

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Features:

  • Floating aluminum heat sink reduces chance of plates cracking during runs.
  • Upper buffer chamber (UBC) requires minimal buffer and is easily drained via inline drain port into removable bottle.
  • Lower buffer chamber (LBC) is removable for simple buffer disposal.
  • Side clamps press gel plates against gasket seated on UBC, applying even pressure across plate and forming a leak-proof seal resulting in flat, even banding patterns.
  • Clamps slide over glass plates for easy setup and removal of gels.

Includes:

  • Removable LBC
  • Heat sink
  • Two side clamps
  • Two blank glass plates
  • Two notched glass plates
  • Two sharktooth combs (78 teeth, 0.4mm thick)
  • Well comb (40 well, 0.4mm thick)
  • Two spacer sets (0.4mm)
  • Adhesive temperature indicator
  • UBC with removable drain bottle (with safety interlocking lid and attached power supply lead)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel: 20 x 45cm; 950mL; 27 x 23 x 50cm
Dimensions (L x W x H) Exterior 10.5 x 9 x 19.5 in. (27 x 23 x 50cm)
Format 40-well
Height (English) Exterior 19.5 in.
Height (Metric) Exterior 50 cm
Length (English) Exterior 10.5 in.
Length (English) Gel 17.716 in.
Length (Metric) Exterior 27 cm
Length (Metric) Gel 45 cm
Volume (English) Buffer 27 fl oz.
Volume (Metric) Buffer 800mL
Width (English) Exterior 9 in.
Width (English) Gel 7.87 in.
Width (Metric) Exterior 23 cm
Width (Metric) Gel 20 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.