Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System
GSA_VA

Catalog No. p-4528895
Click to view available options
Type:
Wide
Wide w/Buffer Exchange Ports

Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.

Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.

  • Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
  • System is excellent for screening PCR samples or routine DNA fragment analysis.
  • Small footprint saves space on lab bench.
  • Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Electrode placement allows gel to be run quickly without compromising resolution.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed UVT gel tray
  • External multiple gel caster
  • Four combs (two 30-well and two 40-well, 1.5mm thick)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Complete System with Buffer Exchange Ports
Product Line Owl
Type Wide w/Buffer Exchange Ports
Sample Size 10 to 80
Volume (Metric) Buffer 600 mL
Dimensions (L x W x H) 7.38 x 7.75 x 4 in. (19 x 20 x 10 cm)
Length (English) 7.38 in.
Length (Metric) 19 cm
Length (Metric) Gel 14.4 cm
Width (English) 7.75 in.
Width (English) Gel 6.5
Width (Metric) 20 cm
Width (Metric) Gel 10.2 cm
Height (English) 4 in.
Height (Metric) 10 cm
Max. Voltage 150 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.