Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat Nos. B1-Rl-9C, -9D, -17C and -17D are 8 and 12-channel pipette format (1X) and (2X) combs.
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.

Specifications
Specifications
Description | Rapid-load 9 tooth (1X) comb; 1.0mm |
For Use With (Equipment) | Owl™ EasyCast B1 Mini Gel Electrophoresis System |
Gel Size | Mini |
Product Line | Owl, EasyCast |
Type | Rapid Load Comb |
Teeth Count | 9 (1X) |
Thickness (Metric) Teeth | 1 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.