Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
GSA_VA

Catalog No. p-6013222
Click to view available options
Includes:
Buffer Exchange Ports
Without Buffer Exchange Ports

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.

Features:

  • System offers a wide range of combs.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.

Includes:

  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
  • Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick

Ordering Information:

Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.

Warranty and Service Offering:

  • Three years

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Includes Buffer Exchange Ports
Sample Size 5 to 34
Volume (Metric) Buffer 600 mL
Dimensions (L x W x H) 8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm)
Length (English) 8.63 in.
Length (Metric) 22 cm
Length (Metric) Gel 11 cm
Width (English) 5.88 in.
Width (Metric) 15 cm
Width (Metric) Gel 9 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.