Learn More
Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.
$2343.09
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Capacity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Capacity | Price | Quantity & Availability | ||||
OWHEP1
|
Thermo Scientific™
HEP1 |
25 x 25 x 5 cm | Transfer area: 20 x 20 cm |
Each of 1 for $2,343.09
|
|
||||
Description
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.
- Fast, efficient transfer of proteins and nucleic acids to membranes
- Buffer contained within blotting paper is sufficient for efficient transfer of molecules
- Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate
- Ideal for proteins less than 150 kDa
Includes:
- Base and lid
- Power supply leads
- Heavy-duty knobs
Specifications
Owl | |
Electroblotting System |