Learn More
OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | OX40 Ligand/TNFSF4 |
| Applications | Blocking Assay, CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Clone | R4930 (Oxelumab) |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Gene Alias | CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 |
| Gene Symbols | TNFSF4 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.