Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OX40/TNFRSF4 Antibody (7D6), Allophycocyanin/Cy7
SDP

Catalog No. N1064375AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064375AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064375AP7 Supplier Novus Biologicals Supplier No. NB10064375APCCY7

Mouse Monoclonal Antibody

OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Feline samples. It is validated for Flow Cytometry.

Specifications

Antigen OX40/TNFRSF4
Applications Flow Cytometry
Classification Monoclonal
Clone 7D6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen
Gene Symbols TNFRSF4
Host Species Mouse
Immunogen CHO-derived feline CD134-Fc fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7293.0
Test Specificity NB100-64375 recognizes feline CD134, otherwise known as OX40, a 43kDa type I membrane protein and member of the tumor necrosis factor receptor superfamily, expressed predominantly by CD4+ activated T cells, and a key regulator of T cell-dependent immune responses. CD134 has been identified as the receptor for the OX40 ligand, CD252, expressed by activated B cells, acting as a co-stimulatory signal for the stimulation and secretion of immunoglobulins. CD134 has also been identified as a binding receptor for Feline Immunodeficiency Virus (FIV), acting along with CXC chemokine receptor 4 (CXCR4) to facilitate the entry of the virus into CD4+ primary target cells.
Target Species Feline
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.