Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OX40/TNFRSF4 Antibody (OX-86), HRP, Novus Biologicals™
SDP

Catalog No. NB10064847H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064847H 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064847H Supplier Novus Biologicals Supplier No. NB10064847H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen OX40/TNFRSF4
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-86
Conjugate HRP
Dilution Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen
Gene Symbols TNFRSF4
Host Species Rat
Immunogen CHO cells transfected with murine OX40
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 7293
Test Specificity NB100-64847 recognises murine OX40 (CD134), a cell surface antigen expressed only by activated T lymphocytes.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.