Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXA1L Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB102055 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102055 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102055 Supplier Novus Biologicals Supplier No. NBP297443JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OXA1L Polyclonal specifically detects OXA1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen OXA1L
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50 mM Sodium Borate
Gene Alias hsa, MGC133129, mitochondrial inner membrane protein OXA1L, OXA1, OXA1Hs, OXA1-like protein, oxidase (cytochrome c) assembly 1-like, Oxidase assembly 1-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OXA1L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5018
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.