Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Oxalic Acid, Dihydrate
SCI_ED

Catalog No. s25455 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade - Solid

Catalog No. S25455

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Assay Percent Range >98%
Linear Formula H2C2O4· 2H2O
Formula Weight 126.07
Grade Laboratory
Density 1.653g/mL at 18.5°C
Quantity 500 g
Packaging Lab Grade - Solid
Chemical Name or Material Oxalic Acid, Dihydrate
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.