Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OXF BD 02

Click to view available options
Quantity:
10 mg
50 mg
Description
OXF BD 02 is a selective inhibitor of the first bromodomain of BRD4 (BRD4(1)) (IC50 = 382 nM). Exhibits 2-3-fold selectivity for BRD4(1) over the CBP bromodomain and has little affinity for a range of other bromodomains. Reduces viability of lung adenocarcinoma cell lines and attenuates proliferation of MV-4-11 leukemia cells. Cell permeable.
Specifications
Specifications
Purity | >98% |
Quantity | 50 mg |
Product Type | OXF BD 02 |
Molecular Weight (g/mol) | 295.33 |
Inhibitors | OXF BD 02 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction