Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker RPF Agar Base

Catalog No. cm0961b
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective medium for the isolation and enumeration of coagulase positive staphylococci from food.

The Oxoid Baird-Parker RPF Agar Base is a selective medium for the isolation and enumeration of coagulase positive staphylococci from food.

  • For use with RPF supplement, Part No. SR0122A
  • Do not use Egg Yolk Tellurite Emulsion
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: ISO, NMKL.
Description Baird-Parker Agar Base, RPF
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.1L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.