Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Blood Agar Base No. 2 (Dehydrated)

Catalog No. oxcm0271b
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate and isolate fastidious pathogens and other microorganisms with this highly nutritious medium

Cultivate and isolate fastidious pathogens and other microorganisms with Thermo Scientific™ Blood Agar Base No. 2 (Dehydrated). The specially designed formulation meets the need for a nutritious blood agar which permits maximum recovery of delicate organisms without interfering with their hemolytic reactions. Without any additions, the medium can be used for short-term maintenance of stock cultures. And with the addition of blood, the medium becomes suitable for the determination of the hemolytic reactions which are important diagnostic criteria for many organisms.

An Improved version of Blood Agar Base, the Blood Agar Base No.2 is a highly nutritious medium generally used as a basal medium for preparing blood agar by addition of blood. It can also be used as general purpose media without the addition of blood.

  • Exhibits superior growth promoting properties and enhanced pigment formation in chromogenic bacteria.
  • Supports luxuriant early colonial development of fastidious organisms, particularly streptococci and pneumococci.
  • Allows determination of hemolytic reaction patterns with the addition of defibrinated blood.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Blood Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.5L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.