Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brain Heart Infusion Broth (Dehydrated)

Catalog No. CM1135R
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy

Catalog No. CM1135R

Supplier: Thermo Scientific™ CM1135R

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate streptococci, Neisseria and other fastidious organisms with this highly nutritious medium.

Cultivate streptococci, Neisseria and other fastidious organisms with Thermo Scientific™ Oxoid™ Brain Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms. It is recommended for blood culture work for the isolation and cultivation of pathogenic fungi.

Cultivate a wide variety of organisms with Brain Infusion Broth:

  • Use as a growth and suspension medium for staphylococci when testing coagulase production.
  • Add 10% defibrillated blood for isolation and cultivation of pathogenic fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Brain Heart Infusion Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg
Yield For 67.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.